Learn More
RUBIS High Precision Tweezers
High Precision Tweezers - Antimagnetic/Anti-acid SS
Brand: RUBIS H
Additional Details : Weight : 0.01000kg
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
Specifications
Fine | |
3.54 in. | |
90 mm | |
Each |
Straight, Strong, Sturdy | |
Anti-acid, Anti-magnetic Stainless Steel | |
High Precision Tweezers | |
Non-magnetic, Antacid Treatment |