Learn More
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
88.60€ - 266.00€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376098
|
Thermo Scientific Alfa Aesar
011089.36 |
500 g |
88.60€
500g |
In Stock
Log in to see stock. |
|||||
11366098
|
Thermo Scientific Alfa Aesar
011089.A3 |
2 kg |
266.00€
2kg |
In Stock
Log in to see stock. |
|||||
Description
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.