Learn More
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
88.60€ - 266.00€
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376098
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
011089.36
500 g
88.60€
500g
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11366098
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
011089.A3
2 kg
266.00€
2kg
View alternative products
Save Up to 
»
In Stock
Description
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.