Learn More
De Buyer™ Stainless Steel Pastry Bowls
Brand: De Buyer™ 3250.28
Additional Details : Weight : 0.46000kg
Specifications
Pastry Bowl | |
For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients | |
125 mm | |
Flat Bottom | |
5.5 L |
Stainless Steel | |
Fridge, deep-freezer, blast-chiller and oven | |
Each | |
Sterile |