Learn More
Fisherbrand™ Stainless Steel Manual Decapper for clean room
Allow easy removal of aluminum seals without breakage
Brand:  Fisherbrand™
1026.00 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 11586034
  • 1080.00€ / Each
Estimated Shipment: 31-05-2024
Description
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
Specifications
Specifications
Manual
13mm Decapper