Learn More
Bochem™ Stainless Steel Graduated Buckets
Product Code. 10312911
Change view
Click to view available options
Diameter (Metric) Outer:
280 mm
285 mm
310 mm
Height (Metric):
210 mm
235 mm
305 mm
Unit Size:
Each
Capacity (Metric):
8 L
15 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) unitSize Capacity (Metric)
10312911 280 mm 235 mm Each -
10683511 310 mm 305 mm Each 15 L
10182741 285 mm 210 mm Each 8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10312911 Supplier Bochem™ Supplier No. 8322
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ Stainless Steel Graduated Buckets have one liter graduations.
  • Type 2: without base ring
  • Graduated per 1 liter
  • Lids must be ordered separately
Specifications
Product Type Graduated Bucket
Material Stainless Steel (18/10)
Diameter (Metric) Outer 280 mm
Height (Metric) 235 mm