Learn More
Stainless Steel gauze, 325 mesh woven from 0.036mm (0.0014in) dia wire, Type 316, Thermo Scientific Chemicals
30.20€ - 91.10€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454075
|
Thermo Scientific Alfa Aesar
046715.FL |
75 x 75 mm |
30.20€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
15464075
|
Thermo Scientific Alfa Aesar
046715.GT |
150 x 150 mm |
50.60€
Each |
In Stock
Log in to see stock. |
|||||
15474075
|
Thermo Scientific Alfa Aesar
046715.HB |
300 x 300 mm |
91.10€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.