Learn More
S Murray and Co™ Stainless Steel Forceps
Product Code. 12342158
Change view
Click to view available options
Length (Metric):
110 mm
Tip Style:
Spatulate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style unitSize
12342158 110 mm Spatulate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12342158 Supplier S Murray and Co™ Supplier No. L325/01
  • ONLINE EXCLUSIVE PRICE
    2.00€
    1.72€ / Pack of 1
    Save 0.28€
    14% Off
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Use these forceps for a wide range of applications. S Murray™ Stainless Steel Forceps are made from only high grade stainless steel in accordance to the specifications of ISO 7153-1.
  • Stainless Steel forceps
  • For manipulating slides and cover slips
Specifications
Product Type Cover Slip Forceps
Material Stainless Steel
For Use With (Application) For manipulating slides and cover slips.
Quantity Each
Tip Style Spatulate
Length (Metric) 110 mm