Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1 with certificate
Product Code. 15555129 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
Unit Size:
1 unit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
15555129 1 g to 1 kg 1 unit
15535139 1 g to 5 kg 1 unit
15555119 1 to 100 g 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15555129
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Classification M1
Weight (Metric) 1 g to 1 kg
Mass 1 g to 1 kg