Learn More
Sampling Systems™ Stainless Steel Bucket
Sampling Systems™ Stainless Steel Buckets are made from a heavy gauge 316L Stainless Steel for durability.
Brand: Sampling Systems™ A586-12
Additional Details : Weight : 20.16000kg
Description
- Made from 316L Stainless Steel
- Crevice free interior
- Swing handle
- Flat base
Specifications
12 L | |
Stainless Steel (316L) | |
310 mm | |
Polished |