Learn More
Fisherbrand™ Stainless Steel Blunt End Forceps
Product Code. 11512123 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
115 mm
130 mm
160 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11512123 160 mm Each
11592113 115 mm Each
11502123 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11512123
  • ONLINE EXCLUSIVE PRICE
    23.55€
    15.91€ / Pack of 1
    Save 7.64€
    32% Off
View all promotions
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Blunt End Forceps with excellent non-wetting properties provide easy cleaning and can be steam or chemically sterilized.
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
  • Chemically resistant and non-contaminating
  • Non-protein binding and providing bio-inert surfaces
  • No critical quantities lost due to sticking
  • No build up on surfaces
  • Ideal for use under severe laboratory conditions
  • Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
  • With guide pin
Specifications
Product Type Forceps
Material PTFE-coated Stainless Steel
Height (Metric) 160 mm
No. per Pack Each
Style Serrated Jaws
Tip Style Blunt