Learn More
De Buyer™ Spring Tongs
Grip, move and transfer food using the stainless steel De Buyer™ Spring Tongs
6.84€ - 12.19€
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11997844
|
De Buyer™
4788.24N |
240 mm |
6.84€
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11907854
|
De Buyer™
4788.30N |
300 mm |
9.06€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
11927854
|
De Buyer™
4788.40N |
400 mm |
12.19€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- For general purpose food handling needs
- Clamp INOX
- Stainless Steel
- Three lengths
Specifications
Clamp Inox | |
Stainless Steel | |
Each |