Learn More
Spreader Blue Polystyrene L-Shaped Spreader
Product Code. 12822775
Change view
Click to view available options
Quantity:
100 Bags of 5
25 bags of 20
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12822775 25 bags of 20 Pack of 500
12322048 100 Bags of 5 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12822775 Supplier Microspec Supplier No. E3
  • ONLINE EXCLUSIVE PRICE
    39.99€
    34.42€ / Pack of 500
    Save 5.57€
    13% Off
In Stock
Add to basket
Edge
Add to basket
High impact L-shape disposable bacterial spreaders
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of either 20 or 5 sterile spreaders.
Specifications
Disposable Single-use
Color Blue
Quantity 25 bags of 20
Shape L-Shape
Material Polystyrene
Product Type Spreader
Sterility Sterile
For Use With (Application) High impact Polystyrene disposable bacterial spreaders