Learn More
Witeg™ Sprayer
Product Code. 11867750
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11867750 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11867750 Supplier Witeg™ Supplier No. 2 147 000
  • ONLINE EXCLUSIVE PRICE
    76.00€
    73.40€ / Pack of 1
    Save 2.60€
    3% Off
View all promotions
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Ideal sprayer system for chromatography. Witeg™ Sprayer comes with an Erlenmeyer flask and an aspirator bulb.
  • Ideal for chromatography
  • Includes Erlenmeyer flask (100mL)
  • Includes aspirator bulb and tubing
  • ST 19/26 joint
Specifications
Capacity (Metric) 100 mL
Type Sprayer System
For Use With (Application) Chromatography
Includes Erlenmeyer Flask (100 mL), Aspirator Bulb and Tubing
Standard Taper Joint ST 19/26