Learn More
Manta Diagnostics™ Sponge Swab in Neutralising Buffer
Includes 60 sponge swabs in Neutralising Buffer, 50 cm2, and 60 sterile self seal sample bags with 60 pairs of sterile gloves
Brand: Manta Diagnostics™ T7
Additional Details : Weight : 3.67000kg
Description
Options of small, medium, or large glovesSpecifications
Sterile | |
60 Swabs, 60 grips seal Bags and 60 pairs of Medium gloves | |
1 | |
60 |