Learn More
Manta Diagnostics™ Sponge Swab in Neutralising Buffer
Includes 60 sponge swabs in Neutralising Buffer, 50 cm2, and 60 sterile self seal sample bags with 60 pairs of sterile gloves
Brand:  Manta Diagnostics™ T6
Additional Details : Weight : 3.67000kg
 View more versions of this product
Product Code. 16614842
  • 90.05€ / Pack of 60
Estimated Shipment: 09-09-2024
to see stock.
Add to basket
Description
Description
Options of small, medium, or large gloves
Specifications
Specifications
Room Temperature
60 Swabs and 60 Grip Seal Bags, 60 pairs of Large gloves
60
Sterile
1