Learn More
Epredia™ Specimen Stages for HM 520, 525, 550 and 560
Use Epredia™ Specimen Stages for Epredia™ HM 520, 525, 550 and 560 Series with HM 520, 525, 550 and 560 Cryostats..
51.45€ - 194.00€
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
12077219
|
Epredia™
715640 |
HM 560 Series | 50 x 75mm |
192.00€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
12097219
|
Epredia™
715700 |
HM 550 Series | 20mm |
51.45€
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
12007229
|
Epredia™
715710 |
HM 550 Series | 30mm |
64.75€
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
12023139
|
Epredia™
715230 |
HM 520 and 525 Series | 40mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12057229
|
Epredia™
715760 |
HM 550 Series | 70 x 75mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||