Learn More
Fisherbrand™ Sound Protection Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Product Code. 15636074 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H
Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15636074 Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H Each
15626074 Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H Each
15646074 Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H Each
15656074 Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15636074
  • ONLINE EXCLUSIVE PRICE
    512.00€
    444.00€ / Pack of 1
    Save 68.00€
    13% Off
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
Reduces the noise generated by ultrasonic cleaning units while providing a secure closure. For use with Ultrasonic Water Baths.
  • Hinged design
  • Available in a range of sizes for all X-tra ST models
  • Manufactured from stainless steel
  • Accessory item
Specifications
Product Type Sound Protection Cover
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Style Hinged Lid
For Use With (Application) Noise reduction for ultrasonic cleaning units
Material Stainless Steel