Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Solid Neoprene Rubber Bungs
Fisherbrand™ Solid Neoprene Rubber Bungs
Product Code. 11517113 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
6 mm
7 mm
9 mm
10 mm
11 mm
13 mm
17 mm
19 mm
25 mm
27 mm
33 mm
39 mm
43 mm
49 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
79 mm
86 mm
92 mm
99 mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
8 mm
9 mm
11.5 mm
12.5 mm
14 mm
16 mm
20.3 mm
22.5 mm
28.5 mm
31 mm
38.5 mm
44.5 mm
49 mm
55 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
89 mm
96 mm
102 mm
109 mm
Length (Metric):
12 mm
14 mm
16 mm
20 mm
24 mm
26 mm
28 mm
32 mm
38.5 mm
42 mm
45.5 mm
49 mm
54 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
28 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Diameter (Metric) Top | Length (Metric) | Unit Size |
|---|---|---|---|---|
| 11517113 | 43 mm | 49 mm | 42 mm | Pack of 2 |
| 11547093 | 10 mm | 12.5 mm | 20 mm | Pack of 20 |
| 11587093 | 17 mm | 20.3 mm | 26 mm | Pack of 20 |
| 11557093 | 11 mm | 14 mm | 24 mm | Pack of 20 |
| 11597093 | 19 mm | 22.5 mm | 28 mm | Pack of 10 |
| 11527103 | 25 mm | 28.5 mm | 28 mm | Pack of 10 |
| 11537103 | 27 mm | 31 mm | 32 mm | Pack of 10 |
| 11567103 | 33 mm | 38.5 mm | 38.5 mm | Pack of 5 |
| 11597103 | 39 mm | 44.5 mm | 38.5 mm | Pack of 5 |
| 11537113 | 49 mm | 55 mm | 42 mm | Pack of 2 |
| 11587083 | 4 mm | 5.8 mm | 14 mm | Pack of 50 |
| 11547113 | 53 mm | 59.5 mm | 45.5 mm | Each |
| 11557113 | 57 mm | 64 mm | 49 mm | Each |
| 11567113 | 62 mm | 69 mm | 49 mm | Each |
| 11577113 | 67 mm | 74 mm | 49 mm | Each |
| 11507093 | 6 mm | 8 mm | 16 mm | Pack of 20 |
| 11587113 | 71 mm | 78 mm | 49 mm | Each |
| 11597113 | 75 mm | 85 mm | 54 mm | Each |
| 11517093 | 7 mm | 9 mm | 16 mm | Pack of 20 |
| 11537093 | 9 mm | 11.5 mm | 20 mm | Pack of 20 |
| 11507123 | 79 mm | 89 mm | 54 mm | Each |
| 11517123 | 86 mm | 96 mm | 54 mm | Each |
| 11527123 | 92 mm | 102 mm | 54 mm | Each |
| 11537123 | 99 mm | 109 mm | 54 mm | Each |
| 11577083 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 10587064 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 11567093 | 13 mm | 16 mm | 24 mm | Pack of 20 |
| 10265114 | 13 mm | 16 mm | 24 mm | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
Product Code.
11517113
-
13.65€
10.59€ / Pack of 2
Save 3.06€
22% Off
Estimated Shipment: 18-09-2026
Log in to see stock.
Log in to see stock.
Add to basket
12.97 EUR valid until 2026-08-30
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
Add to basket
Specifications
Specifications
| For Use With (Application) | For laboratory use |
| Hardness | 46/55 IRHD |
| Length (Metric) | 42 mm |
| Quantity | 2/Pk. |
| Color | Gray |
| Diameter (Metric) Bottom | 43 mm |
| Diameter (Metric) Top | 49 mm |
| Material | Neoprene |
| Product Type | Bung |