missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tungstate Dihydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 5 g |
Linear Formula | Na2WO4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 329.85g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Tungstate Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'