Learn More
Rogo Sampaic™ Soda Lime Glass Test Tubes
Product Code. p-8003246
Change view
Click to view available options
Diameter (Metric):
15.3 mm
17.3 mm
Length (Metric):
160 mm
180 mm
Unit Size:
Pack of 250
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) unitSize
11701696 17.3 mm 180 mm Pack of 250
11835953 15.3 mm 160 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11701696 Supplier Rogo Sampaic™ Supplier No. BGB032
Add to basket
Add to basket
Ideal for routine laboratory analysis and procedures. Rogo Sampaic™ Soda Lime Glass Test Tubes are economical while still offering good chemical resistance.
  • Thin wall
  • Available in two different capacities
Specifications
Description Test tube
Length (Metric) 180 mm
Material Soda Lime Glass
Quantity 250 Pack
Diameter (Metric) 17.3 mm