Learn More
Hamilton™ Small Hub Removable Needle Syringe
Used exclusively on RN CTC syringes.
Brand:  Hamilton™ 203481
Additional Details : Weight : 0.00100kg
Product Code. 11513024
  • 32.00€ / Each
Estimated Shipment: 17-06-2024
Description
  • Length of 51mm
  • Point style 3 (blunt)
  • Sold in packs of 3
Specifications
Specifications
Small RN (CTC) hub
22s G
3/Pk.
3