Description
- Impress guests with these effective hand towels, featuring attractive leaf embossing and a decorative Tork logo.
- Boost hygiene by using these hand towels with the one-at-a-time dispensing system of the Tork Singlefold H3 dispenser.
- Upgrade your washroom - these paper hand towels are ideal for spaces with moderate visitors.
- Easily open, fold and transport paper hand towels with our Tork Easy Handling™ cardboard packaging.
Specifications
Specifications
| Product Type | Hand Towel |
| Length (English) | 9.06 in. |
| Length (Metric) | 23 cm |
| Width (English) | 9.76 in. |
| Width (Metric) | 24.8 cm |
| Certifications/Compliance | EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, ISO 9001, ISO 14001, FSC Recycled credit, Blauer Engel |
| Color | Green |
| Ply | 2-ply |
| Certifications/Compliance (Europe) | EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, ISO 9001, ISO 14001, FSC Recycled credit, Blauer Engel |