Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents
Brand:  Masterflex™ 95644-26
Additional Details : Weight : 0.20410kg
 View more versions of this product
Product Code. 16609571
  • 119.00€ / Each
Estimated Shipment: 29-08-2024
Description
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Specifications
Gamma Irradiated
1 L