Learn More
DWK Life Sciences Silicone Sealing Ring, VMQ
Product Code. 11747118 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inside:
11.0 to 13.0 mm
13.0 to 15.0 mm
15.0 to 17.0 mm
17.0 to 19.0 mm
25.0 to 27.0 mm
31.0 to 33.0 mm
5.5 to 6.5 mm
7.5 to 9.0 mm
9.0 to 11.0 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Pack of 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Joint Size Unit Size
11747118 5.5 to 6.5 mm GL 14 Pack of 10
11777118 25.0 to 27.0 mm GL 45 Pack of 10
11831010 11.0 to 13.0 mm GL 25 Pack of 10
11786828 7.5 to 9.0 mm GL 18 Pack of 10
11338803 7.5 to 9.0 mm GL 25 Pack of 10
11757198 9.0 to 11.0 mm GL 25 Pack of 10
11777198 9.0 to 11.0 mm GL 18 Pack of 10
12851054 13.0 to 15.0 mm GL 32 Pack of 10
13157444 15.0 to 17.0 mm GL 32 Pack of 10
13118043 17.0 to 19.0 mm GL 32 Pack of 10
11331485 31.0 to 33.0 mm GL 45 Pack of 10
11820720 5.5 to 6.5 mm GL 18 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11747118 Supplier DWK Life Sciences Supplier No. 292340606
  • ONLINE EXCLUSIVE PRICE
    47.36€
    42.57€ / Pack of 10
    Save 4.79€
    10% Off
In Stock
Add to basket
Edge
Add to basket
The silicone seal is used for sealing and fixing threaded pipes with threaded caps.
The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.
  • With bonded PTFE face
Specifications
Diameter (Metric) Inside 5.5 to 6.5 mm
For Use With (Application) Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture
Joint Size GL 14
Quantity 10/Pk.
Size (Metric) Fits Tubing ID: 5.5 to 6.5 mm
Diameter (Metric) Outside 12 mm
Material Silicone (VMQ)