Learn More
Witeg™ Short Stem Funnel
Transfer or filter material with ease. Witeg™Short Stem Funnel is durable and resistant.
Brand:  Witeg™ 5 550 070 B
Additional Details : Weight : 0.80000kg
Product Code. 11877850
  • 40.75€ / Pack of 10
Estimated Shipment: 26-06-2024
Description
  • Short stem: 70mm.
  • Stem O.D.: 8mm.
  • Funnel
  • Borosilicate 3.3.
  • DIN 12445
Specifications
Specifications
70mm
Glass
70mm
8mm
DIN 12445