Learn More
Fisherbrand™ Sheet Compressed Cork
Product Code. 12396447 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Material:
Cork
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12396447 Cork Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12396447
  • ONLINE EXCLUSIVE PRICE
    35.78€
    25.46€ / Pack of 5
    Save 10.32€
    28% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Suitable for entomological work.
Compressed sheet with thickness of 6mm
Specifications
For Use With (Application) For Entomological Work
Quantity 5/Pk.
Size 300 x 300mm
Material Cork
Product Type Insect Spreading Board