Learn More
Epredia™ Shandon™ ColorFrost™ Plus Slides
Epredia™ Shandon ColorFrost Plus Slides are positively charged for improved cellular adhesion.
55.75€ - 963.10€
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
12627976
|
Epredia™
6776214 |
White | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12608106
|
Epredia™
6776214CS |
White | 1440/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12668016
|
Epredia™
9991001 |
Blue | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12678016
|
Epredia™
9991002 |
Yellow | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12688016
|
Epredia™
9991003 |
Green | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12698016
|
Epredia™
9991004 |
Pink | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12608026
|
Epredia™
9991009 |
Lavender | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12618026
|
Epredia™
9991011 |
Burnt Orange | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12628026
|
Epredia™
9991012 |
Tan | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12638026
|
Epredia™
9991013 |
Aqua | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12648026
|
Epredia™
9991014 |
Gold | 100/Cs. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Electrostatically attract tissue sections and cytology preparations, binding them to the slide
- Eliminate the need for special adhesives or protein coatings
- Avoid background staining
- Available in a variety of colors
Specifications
Yes | |
Microscope Slide |