Learn More
Trajan™ Diamond Autosampler Syringes for Agilent and Shimadzu GC
For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan™ 0355411
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13177926
  • 98.10€ / Each
Estimated Shipment: 12-06-2024
Description
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Specifications
Needle coating: Ultra Smooth Hydrophilic
0.004 in.
0.024 in.
Gas chromatography
GC Autosampler Syringe
42 mm
Agilent and Shimadzu
23 G
0.11 mm
0.63 mm
1.65 in.
Ea.
Cone