Learn More
Trajan™ Autosampler Syringes With Removable Needle for Thermo Scientific GCs
For reduced non specific sample interaction and increased analyte recovery
89.90€ - 173.00€
Specifications
For Use With (Application) | Gas chromatography |
---|---|
Product Type | Autosampler Syringe |
Quantity | Ea. |
Needle Point Style | Cone |
For Use With (Equipment) | Thermo GC, CTC PAL RTC (Robotic Tool Change) Autosampler |
Product Code | Brand | Diameter (English) Outer Needle | Needle Gauge | Length (English) Needle | Model | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outer Needle | Needle Gauge | Length (English) Needle | Model | Price | Quantity & Availability | ||
13169195
|
Trajan™
000680 |
0.024 in. | 23 G | 2.24 in. | 1BR-RTC/RSH-5.7/0.63C |
131.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13159015
|
Trajan™
001877 |
0.18 in. | 26 G | 3.34 in. | 5R-RTC/RSH-8.5/0.47C |
173.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13159195
|
Trajan™
000480 |
0.024 in. | 23 G | 2.24 in. | 0.5BR-RTC/RSH-5.7/0.63C |
146.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13126115
|
Trajan™
001871 |
0.024 in. | 23 G | 2.24 in. | 5R-RTC/RSH-5.7/0.63C |
125.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13149205
|
Trajan™
002878 |
0.18 in. | 26 G | 3.34 in. | 10R-RTC/RSH-GT-8.5/0.47C |
140.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13149015
|
Trajan™
001875 |
0.18 in. | 26 G | 2.24 in. | 5R-RTC/RSH-5.7/0.47C |
125.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13129205
|
Trajan™
002874 |
0.24 in. | 23 G | 3.34 in. | 10R-RTC/RSH-GT-8.5/0.63C |
140.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13119205
|
Trajan™
002872 |
0.24 in. | 23 G | 2.24 in. | 10R-RTC/RSH-GT-5.7/0.63C |
115.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13129025
|
Trajan™
002875 |
0.18 in. | 26 G | 2.24 in. | 10R-RTC/RSH-5.7/0.47C |
93.60€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13109025
|
Trajan™
002871 |
0.24 in. | 23 G | 2.24 in. | 10R-RTC/RSH-5.7/0.63C |
89.90€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
13139205
|
Trajan™
002876 |
0.18 in. | 26 G | 2.24 in. | 10R-RTC/RSH-GT-5.7/0.47C |
115.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
Description
- Borosilicate glass barrel for strength and inertness.
- Stainless steel needles for inertness, strength and corrosion resistance.
- Backing behind the scale aids quick and accurate reading of the syringe scale.
- Metal back flange provides stability and resists fracture.
- Metal needle cap on syringe barrel protects and improves syringe lifetime.
- Syringes are individually tested for quality ensuring reliability and conformance.
- Unique packaging design eliminates possible contamination in the syringe.
Specifications
Gas chromatography | |
Ea. | |
Thermo GC, CTC PAL RTC (Robotic Tool Change) Autosampler |
Autosampler Syringe | |
Cone | |
Removable |