Learn More
- Home
- Products
- Vials
- Autosampler Vials, Inserts, and Closures
- Chromatography Autosampler Vial Septa
- Fisherbrand™ Septum for 8-425 screw thread cap
Fisherbrand™ Septum for 8-425 screw thread cap
Product Code. 11787557 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Beige, Red
Beige, Red-Orange
Black
Blue Transparent, White
Blue, White
Cream, Red
Dark Blue, White
Gray, Red
Red, White
Red-Orange, Transparent
White
Material:
Butyl, PTFE
Natural Rubber, TEF
PTFE
PTFE Virginal
PTFE, Rubber
PTFE, Silicone
Viton 1A
Thickness (Metric):
0.25 mm
0.9 mm
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 100
Pack of 1000
15 product options available for selection
| Product Code. | Color | Material | Thickness (Metric) | Unit Size |
|---|---|---|---|---|
| 11787557 | Red-Orange, Transparent | Natural Rubber, TEF | 1.3 mm | Pack of 1000 |
| 11841653 | Blue, White | PTFE, Silicone | 0.9 mm | Pack of 1000 |
| 11872663 | Red, White | PTFE, Silicone | 1.3 mm | Pack of 1000 |
| 11850772 | Blue Transparent, White | PTFE, Silicone | 1.3 mm | Pack of 1000 |
| 11821663 | White | PTFE Virginal | 0.25 mm | Pack of 1000 |
| 11828961 | Red, White | PTFE, Silicone | 1 mm | Pack of 1000 |
| 11830772 | Cream, Red | PTFE, Silicone | 1.5 mm | Pack of 1000 |
| 12950841 | Beige, Red-Orange | PTFE | 1 mm | Pack of 100 |
| 16160961 | Beige, Red-Orange | PTFE | 1 mm | Pack of 1000 |
| 11858961 | Black | Viton 1A | 1 mm | Pack of 1000 |
| 11838961 | Black | Viton 1A | 1.5 mm | Pack of 1000 |
| 11840772 | Dark Blue, White | PTFE, Silicone | 1.3 mm | Pack of 1000 |
| 11868961 | Gray, Red | Butyl, PTFE | 1.3 mm | Pack of 1000 |
| 10258004 | Beige, Red | PTFE, Rubber | 1 mm | Pack of 1000 |
| 11848961 | Red-Orange, Transparent | Natural Rubber, TEF | 1 mm | Pack of 1000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
Product Code.
11787557
Big Savings for Better Separation
Use promo code 27992
Use promo code 27992
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
77.24 EUR valid until 2026-10-31
BEST PRICE on Promo! Use promo code "27992" to get your promotional price.
BEST PRICE on Promo! Use promo code "27992" to get your promotional price.
Add to basket
Specifications
Specifications
| Description | Septa |
| Color | Red-Orange, Transparent |
| Diameter (Metric) Outer | 8 mm |
| Material | Natural Rubber, TEF |
| Quantity | 1000 |
| Thickness (Metric) | 1.3 mm |
| Product Type | 1.3 mm |