Learn More
Witeg™ Separatory Funnel, Pear-Shaped
Use this funnel to separate the components of a liquid-liquid mixture. Witeg™ Separatory Funnel, Pear-Shaped is designed with resistant materials.
Brand: Witeg™ 2 647 100
Additional Details : Weight : 0.80000kg
Description
- Ungraduated.
- Pear-shaped.
- Separates liquid mixtures.
Specifications
100 mL | |
Bore size 2.5mm | |
ST 19/26 |
Borosilicate Glass | |
No | |
ST PE 19/26 |