Learn More
DWK Life Sciences Wheaton™ Screw Caps For Wheaton™ E-Z Vials™
For 12mm x 32mm E-Z Vials™
174.00€ - 194.00€
Specifications
Material (Liner) | Linerless |
---|---|
Color | Black |
Material | Polypropylene |
Closure Size | 10-425 |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12645010
|
DWK Life Sciences
242765 |
Without top seal |
174.00€
Pack of 1000 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
12498862
|
DWK Life Sciences
242766 |
With top seal |
194.00€
Pack of 1000 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
Description
- Screw Cap
- Polypropylene
- 10mm (0.39in.)
- Unlined
- With solid top
- Black
Specifications
Linerless | |
Black | |
Polypropylene | |
10-425 |