Learn More
Labcon™ Screw Caps with Elastomeric Seal for SuperClear™ Microtubes
These screw caps with elastomeric seals are available in a wide range of colors.
Brand: Labcon™ 3602-878-000-9
Additional Details : Weight : 0.21540kg
Description
- For SuperClear™ microcentrifuge tubes
- With integral elastomeric seal
- Cap includes soft gripping surface
- Endotoxin-free (non-pyrogenic)
- Polyethylene USP Class VI resin
- Made in USA
Specifications
12.7 mm | |
Green | |
6.1 mm | |
5000/Cs. | |
Non-sterile |
Polypropylene | |
Bagged | |
500/Pk. | |
500 Caps/Pk., 10 Pk./Cs. | |
0.5 to 2 mL |