Learn More
Sartorius Vivacon™ 2 DNA Concentrator
Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples
Brand: Sartorius VN02H01
Additional Details : Weight : 0.10000kg
Description
Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.
- Convenient handling in fixed-angle rotors
- Low binding material
- Reverse spin possibility
- Graduation and MWCO printed on device
- Complete and highly reproducible concentrate recovery
Specifications
50ng/mL | |
Polycarbonate Body, Polypropylene Filtrate Vessel, Polypropylene Back Spin Vial, Polypropylene Concentrator Cap, Hydrosart Membrane, Silicone O-ring | |
0.95 cm2 | |
5000xG | |
DNA, peptide | |
2 mL | |
4.92 x 0.62 in. (12.5 x 1.6cm) | |
4.52 in. | |
11.5 cm | |
1.6 cm |
60 min. | |
10 kDa | |
DNA concentration, desalting, buffer exchange, fractionation | |
25 Pack | |
Vivacon 2 DNA Concentrator | |
10 μL | |
4.92 in. | |
12.5 cm | |
0.62 in. |