Learn More
Sartorius Sterisart™ NF Sterility Testing System
BIOPHARMA
Product Code. p-4231981 Shop All Sartorius Products
Change view
Click to view available options
Sterilization Method:
Gamma Irradiation
Includes:
Long dual-needle spike, sterile vented
Long dual-needle spike, sterile vented, septum
Long needle, by-packed sterile venting needle
Long needle, sterile venting needle, septum
Short dual-needle spike, sterile vented, with septum
Tubing system w/ two dual-needle metal spikes of different length
Universal Luer adapter, long dual-needle spike, sterile vented, septum
Unit Size:
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterilization Method Includes Unit Size
11700965 Gamma Irradiation Long dual-needle spike, sterile vented, septum Pack of 10
11790955 Gamma Irradiation Long dual-needle spike, sterile vented Pack of 10
11720965 Gamma Irradiation Long needle, by-packed sterile venting needle Pack of 10
11770965 Gamma Irradiation Tubing system w/ two dual-needle metal spikes of different length Pack of 10
11730965 Gamma Irradiation Long needle, sterile venting needle, septum Pack of 10
15972530 Gamma Irradiation Short dual-needle spike, sterile vented, with septum Pack of 10
11760965 Gamma Irradiation Universal Luer adapter, long dual-needle spike, sterile vented, septum Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11700965 Supplier Sartorius Supplier No. 16466GSD
Add to basket
Add to basket
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius Sterisart™ NF Sterility Testing System is optimal for use with isolators.
International pharmacopoeias require the complete sterility of pharmaceutical products that are injected into the blood stream or that otherwise enter the body below the skin surface. As a manufacturer of such products, you are required to supply proof of sterility of the final product batch. Sterisart™ NF is a completely closed system for the sterility testing of pharmaceutical products. It is based on the membrane filter method, however it eliminates the procedure of manipulating the filters. By this the main risk of a secondary contamination and false positive results is eliminated. A peristaltic pump transfers the sample into the filtration units, and after rinsing, the filtration units are filled with media and used for incubation of the filters without any contact to the environment.
  • reliable Sartochem™ membrane
  • high retention of microbes
  • low adsorption
  • high mechanical stability
  • pre-installed color-coded tube clamps
  • easy-to-read graduated marks
  • user-friendly (several practical adapters available)
  • product-/lot number identification
  • gas-impermeable packaging for protection against sterilants
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 0.45 μm
Sterility Sterile
Filtration Area 15.7 cm
For Use With (Application) Sterility Testing
Includes Long dual-needle spike, sterile vented, septum
Max. Pressure 3 bar at 20°C
Sample Type LVPs
Sterilization Method Gamma Irradiation
Temperature (English) Max. 122°F
Temperature (Metric) Max 50°C
Volume (English) 4 oz.
Volume (Metric) 120 mL
Closure Type Septum