Learn More
Sartorius Stainless Steel Pressure Filter Holder
16249 is a practical filter holder for the filtration of up to 5 liter volumes. It can be attached to a tripod with the help of a steel rod which can be screwed in.
Brand:  Sartorius 16249
Additional Details : Weight : 1.19700kg
Product Code. 15992520
  • 1645.00€ / Each
Estimated Shipment: 16-05-2024
Description
16249 is a practical filter holder for the filtration of up to 5 liter volumes. It can be attached to a tripod with the help of a steel rod which can be screwed in.
  • 16249 is a practical filter holder for the filtration of up to 5 liter volumes
  • It can be attached to a tripod with the help of a steel rod which can be screwed in
Specifications
Specifications
Stainless Steel
13 cm2
Pressure Filter Holder
200 mL
4.7 cm
Stainless Steel
Each
200 mL
200 mL