Learn More
Sartorius Quantitative Grade 389 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
Brand:  Sartorius FT-3-102-100
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 15902850
  • 35.50€ / Pack of 100
Estimated Shipment: 05-06-2024
Description
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
White
8 to 12 μm
389
0.19 mm
Disc
20 sec.
100/Pk.
100 mm