Learn More
Sartorius Printer Paper
Product Code. 10663182 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
YDP06 Dot Matrix Printer
YDP20-0CE Printer
Length (English):
550 in.
1944 in.
Length (Metric):
1397 cm
4937.76 cm
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Length (English) Length (Metric) Unit Size
10663182 YDP20-0CE Printer 1944 in. 4937.76 cm Pack of 5
11338963 YDP06 Dot Matrix Printer 550 in. 1397 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10663182 Supplier Sartorius Supplier No. 6906937
  • ONLINE EXCLUSIVE PRICE
    27.60€
    26.10€ / Pack of 5
    Save 1.50€
    5% Off
In Stock
Add to basket
Edge
Add to basket
Paper for various Sartorius printers.
Paper for use with balance printers.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) YDP20-0CE Printer
Length (English) 1944 in.
Length (Metric) 4937.76 cm
Type Printer Paper