Learn More
Sartorius Entris™ Toploading Balances
When you need to get a heavy workload of repetitive applications done fast. Innocal Calibration Services avaliable on some models.
941.00€ - 1700.00€
Specifications
Connection Type | RS-232C, 25-pin |
---|---|
Stabilization Time | 1.5 sec. |
Type | Toploading Balance |
Product Code | Brand | Capacity | Readability | Dimensions (L x W x H) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Dimensions (L x W x H) | Includes | Price | Quantity & Availability | ||
15203923
|
Sartorius
ENTRIS6202-1S |
6.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15213923
|
Sartorius
ENTRIS4202-1S |
4.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15223923
|
Sartorius
ENTRIS3202-1S |
3.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15233923
|
Sartorius
ENTRIS2202-1S |
2.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15243923
|
Sartorius
ENTRIS822-1S |
820 g | 10 mg | 303 x 230 x 87 mm (12.9 x 9.0 x 3.4 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15253923
|
Sartorius
ENTRIS8201-1S |
8.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15263923
|
Sartorius
ENTRIS5201-1S |
5.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15273923
|
Sartorius
ENTRIS2201-1S |
2.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | External Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15293903
|
Sartorius
ENTRIS822I-1S |
820 g | 10 mg | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
15283903
|
Sartorius
ENTRIS2202I-1S |
2.2 kg | 10 mg | 303 x 230 x 91 cm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15223913
|
Sartorius
ENTRIS2201I-1S |
2.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15203913
|
Sartorius
ENTRIS8201I-1S |
8.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15263903
|
Sartorius
ENTRIS4202I-1S |
4.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15234946
|
Sartorius
ENTRIS5201I-1S |
5.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15213913
|
Sartorius
ENTRIS5201I-1S |
5.2 kg | 100 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
15273903
|
Sartorius
ENTRIS3202I-1S |
3.2 kg | 10 mg | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) | Internal Calibration |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
Description
- White-backlit, high-contrast display with 15mm digits; exceptionally easy to read under any room lighting conditions
- New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
- RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
- Total ease of operation
- All keys have positive click action
- Clearly defined control panel
- Monolithic weighing cell provides high thermal stability and extremely fast response times
- Fast response time and display stability
- Language-driven user interface
- Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Innocal Calibration Services
Available in items containing "i-1SUS" in Manufacturer No.: 14-559-023, -024,- 025, -026, -027, -028, -029, -030.
Specifications
RS-232C, 25-pin | |
1.5 sec. | |
Toploading Balance |