Learn More
Sartorius Accessories for Combisart™ Systems: Forceps
Forceps for use with Sartoris Combisart Systems
Brand: Sartorius 16625
Additional Details : Weight : 0.25000kg
Description
- Stainless Steel
- Blunt tip
- Can be flamed, also autoclavable
Specifications
Tweezers | |
Firm hold of the membrane filter | |
Blunt-edged tips |
Stainless Steel | |
Vacuum filter holders and manifold system |