Learn More
Sartorius Qualitative Grade 601/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Brand:  Sartorius FT-3-354-185
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11791335
  • 19.50€ / Pack of 100
Estimated Shipment: 11-06-2024
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifications
Specifications
Disc
601/N
0.19 mm
13 sec.
50/Pk.
185 mm