Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand:  Sartorius FT-3-215-185
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10495981
  • 40.30€ / Pack of 100
Estimated Shipment: 11-06-2024
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
Disc
97 sec.
100/Pk.
185 mm
4 to 7 μm
292 a
0.19 mm