Learn More
Sartorius™ Sartopore™ 2 MidiCaps Polyethersulfone Membrane Filter Capsules
Brand:  Sartorius™ 5445307G7--FF--A
Additional Details : Weight : 0.50000kg
Product Code. 15288073
  • 925.38€ / Pack of 4
Estimated Shipment: 10-10-2024
Specifications
Autoclavable
PES
Sterile
0.75 in. Tri-Clamp
0.75 in. Tri-Clamp
Filter Capsule
ISO 9001, USP Class VI, cGMP Standard
0.8/0.2 μm
0.065 m2
4Pack
4Pack
77 mm