Learn More
Fisherbrand™ Sample Storage Trays
Product Code. 15376548 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
White, Blue
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15376548 White, Blue Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15376548
  • ONLINE EXCLUSIVE PRICE
    123.00€
    108.57€ / Pack of 10
    Save 14.43€
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Separate items with included dividers or remove them for general product storage and transport.
  • Made from corrugated polypropylene
  • Moisture resistant, significantly reducing the chance of mold
  • Lightweight and sturdy to hold up to 24 full specimen containers
  • Stackable with other trays
  • Removable dividers allow outer shell to be used for storage or transport of laboratory products
  • Supplied flat-packed
  • Pack includes: 10 trays and 10 dividers and 5/CS
  • Disposable
Specifications
Product Type Storage Tray
Color White, Blue
Material Corrugated Polypropylene
Dimensions (L x W x H) 406 x 260 x 89 mm
Includes Removable insert
Insert Type 24-well divider