Learn More
R&L Enterprises™ Bright Stainless Steel Rods
Product Code. 11785386 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
Diameter (Metric) Inner:
10 mm
12.5 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Unit Size
11785386 500 mm 10 mm Each
11795386 750 mm 12.5 mm Each
11759327 500 mm 12.5 mm Each
11705396 1000 mm 12.5 mm Each
12328149 600 mm 12.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11785386
In Stock
Add to basket
Edge
Add to basket
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Bright Stainless Steel Rods are durable and corrosion resistant.
  • Threaded
  • Length of rod does not include the thread
  • Durable
  • Easy-to-clean
  • Corrosion resistant
  • Stainless steel
Specifications
Material Stainless Steel
Length (Metric) 500 mm
Diameter (Metric) Inner 10 mm
Product Type Rod (Threaded)
Description Threaded
For Use With (Application) For Rectangular Stand