Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Solid Rubber Stoppers
Fisherbrand™ Solid Rubber Stoppers
Product Code. 11512922 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
5 mm
7 mm
8 mm
9 mm
10 mm
13 mm
15 mm
19 mm
21 mm
27 mm
29 mm
31 mm
33 mm
37 mm
39 mm
41 mm
43 mm
45 mm
49 mm
53 mm
71 mm
75 mm
86 mm
99 mm
Stopper Number:
3
4
5
7
8
9
10
13
15
19
21
27
29
31
33
37
39
41
43
45
49
53
71
75
86
99
Height (Metric):
12mm
14mm
16mm
20mm
24mm
28mm
32mm
35mm
38.5mm
42mm
45.5mm
49mm
54mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
7 mm
9 mm
10.5 mm
11.5 mm
12.5 mm
16 mm
18 mm
22.5 mm
24.5 mm
31 mm
33 mm
36 mm
38.5 mm
42.5 mm
44.5 mm
47 mm
49 mm
51 mm
55 mm
59.5 mm
78 mm
85 mm
96 mm
109 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
27 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Stopper Number | Height (Metric) | Diameter (Metric) Top | unitSize |
|---|---|---|---|---|---|
| 11512922 | 10 mm | 10 | 20mm | 12.5 mm | Pack of 20 |
| 11532922 | 13 mm | 13 | 24mm | 16 mm | Pack of 20 |
| 11542922 | 15 mm | 15 | 24mm | 18 mm | Pack of 20 |
| 11562922 | 19 mm | 19 | 28mm | 22.5 mm | Pack of 20 |
| 11572922 | 21 mm | 21 | 28mm | 24.5 mm | Pack of 20 |
| 11502932 | 27 mm | 27 | 32mm | 31 mm | Pack of 10 |
| 11512932 | 29 mm | 29 | 32mm | 33 mm | Pack of 10 |
| 10355351 | 31 mm | 31 | 35mm | 36 mm | Pack of 10 |
| 11532932 | 33 mm | 33 | 38.5mm | 38.5 mm | Pack of 10 |
| 11552932 | 37 mm | 37 | 38.5mm | 42.5 mm | Pack of 10 |
| 11562932 | 39 mm | 39 | 38.5mm | 44.5 mm | Pack of 10 |
| 11542912 | 3 mm | 3 | 12mm | 4.5 mm | Pack of 50 |
| 10558431 | 41 mm | 41 | 42mm | 47 mm | Pack of 5 |
| 11582932 | 43 mm | 43 | 42mm | 49 mm | Pack of 5 |
| 10191411 | 43 mm | 43 | 42mm | 49 mm | Pack of 5 |
| 11502942 | 49 mm | 49 | 42mm | 55 mm | Pack of 5 |
| 11592932 | 45 mm | 45 | 42mm | 51 mm | Pack of 5 |
| 11552912 | 4 mm | 4 | 14mm | 5.8 mm | Pack of 50 |
| 11512942 | 53 mm | 53 | 45.5mm | 59.5 mm | Pack of 2 |
| 11562912 | 5 mm | 5 | 16mm | 7 mm | Pack of 50 |
| 11557582 | 71 mm | 71 | 49mm | 78 mm | Each |
| 11587582 | 86 mm | 86 | 54mm | 96 mm | Each |
| 11567582 | 75 mm | 75 | 54mm | 85 mm | Each |
| 11582912 | 7 mm | 7 | 16mm | 9 mm | Pack of 20 |
| 11592912 | 8 mm | 8 | 20mm | 10.5 mm | Pack of 20 |
| 11507592 | 99 mm | 99 | 54mm | 109 mm | Each |
| 10151801 | 9 mm | 9 | 20mm | 11.5 mm | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
Artikelnummer.
11512922
-
7.35€
5.46€ / Pack of 20
Save 1.89€
25% Off
Estimated Shipment: 31-08-2026
Log in to see stock.
Log in to see stock.
Læg i indkøbskurv
6.98 EUR valid until 2026-08-30
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
Læg i indkøbskurv
Tekniske data
Tekniske data
| For Use With (Application) | Laboratory use |
| Height (Metric) | 20mm |
| Quantity | 20/Pk. |
| Color | Red |
| Diameter (Metric) Bottom | 10 mm |
| Diameter (Metric) Top | 12.5 mm |
| Material | Rubber |
| Product Type | Solid Stopper |
| Stopper Number | 10 |
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?