Learn More
- Home
- Products
- Stoppers and Closures
- Corks
- Fisherbrand™ Cork Stoppers
Fisherbrand™ Cork Stoppers
Product Code. 12346457 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
6 mm
8 mm
9 mm
12 mm
13 mm
14 mm
17 mm
19 mm
21 mm
22 mm
25.5 mm
28.5 mm
30 mm
32 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
Diameter (Metric) Top:
9.5 mm
11 mm
12 mm
15 mm
16 mm
17 mm
20.5 mm
22 mm
24 mm
25.5 mm
28.5 mm
32 mm
33 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
57 mm
Unit Size:
Pack of 10
Pack of 20
21 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Diameter (Metric) Top | unitSize |
|---|---|---|---|
| 12346457 | 12 mm | 15 mm | Pack of 20 |
| 12356457 | 13 mm | 16 mm | Pack of 20 |
| 12366457 | 14 mm | 17 mm | Pack of 20 |
| 12386457 | 17 mm | 20.5 mm | Pack of 10 |
| 12316467 | 21 mm | 24 mm | Pack of 10 |
| 12336467 | 25.5 mm | 28.5 mm | Pack of 10 |
| 12326467 | 22 mm | 25.5 mm | Pack of 10 |
| 12306467 | 19 mm | 22 mm | Pack of 10 |
| 12346467 | 28.5 mm | 32 mm | Pack of 10 |
| 12356467 | 30 mm | 33 mm | Pack of 10 |
| 12803834 | 30 mm | 33 mm | Pack of 10 |
| 12366467 | 32 mm | 35 mm | Pack of 10 |
| 12376467 | 35 mm | 38 mm | Pack of 10 |
| 12386467 | 38 mm | 41.5 mm | Pack of 10 |
| 12396467 | 41.5 mm | 44 mm | Pack of 10 |
| 12306477 | 44 mm | 47.5 mm | Pack of 10 |
| 12316477 | 47.5 mm | 50 mm | Pack of 10 |
| 12306457 | 6 mm | 9.5 mm | Pack of 20 |
| 12326477 | 50 mm | 57 mm | Pack of 10 |
| 12316457 | 8 mm | 11 mm | Pack of 20 |
| 12326457 | 9 mm | 12 mm | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
Product Code.
12346457
-
3.15€
2.65€ / Pack of 20
Save 0.50€
15% Off
Estimated Shipment: 31-08-2026
Log in to see stock.
Log in to see stock.
Add to basket
2.99 EUR valid until 2026-08-30
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
Add to basket
Specifications
Specifications
| Length (Metric) | 21 mm |
| Quantity | 20/Pk. |
| Diameter (Metric) Bottom | 12 mm |
| Diameter (Metric) Top | 15 mm |
| Material | Cork |
| Product Type | Cork Stopper |