Learn More
Abeba™ Rubber 5150 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5150/40
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15651828
  • 70.94€ / 1 pair
Estimated Shipment: 04-06-2024
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Microfiber
40
Rubber
Blue
Leather
Slip-resistant
5150