Learn More
Lenz Laborglasintrumente Round Bottom Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07035079
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10356361
  • 172.78€ / Each
Estimated Shipment: 11-07-2024
Description
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
76 mm
4 L
Duran Glass
290 mm