Learn More
Lenz Laborglasintrumente Round Bottom Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07035049
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10336361
  • 345.56€ / Pack of 10
Estimated Shipment: 17-07-2024
Description
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
51 mm
250 mL
Duran Glass
143 mm